Skip to Content
Merck
All Photos(1)

Documents

U6135

Sigma-Aldrich

Ubiquitin-Specific Protease 8 human

recombinant, expressed in Sf9 cells, ≥70% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.77

recombinant

expressed in Sf9 cells

Quality Level

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

128 kDa

concentration

≥0.10 mg/mL protein (Bradford)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

General description

Human USP8 (Ubiquitin-Specific Protease 8) or UBPY, amino-acids 2-1118 (end) with an N-terminal FLAG-tag, expressed in a baculovirus-infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Expression of ubiquitin-specific protease 8 (USP8) in mice is critical for proper expression and stability of several growth factor receptors including epidermal growth factor receptor. However, USP8 was shown to degrade chemokine receptor 4.

Physical form

Solution in 45 mM Tris-HCl, 124 mM NaCl, 2.4 mM KCl, 90 ng/μl Flag peptide, 3 mM DTT, and 20% Glycerol at pH 8.0.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essential Role of Ubiquitin-Specific Protease 8 for Receptor Tyrosine Kinase Stability and Endocytic Trafficking In Vivo
Niendorf, S., et al.
Molecular and Cellular Biology, 727, 5029-5039 (2007)
The Deubiquitinating Enzyme USP8 Promotes Trafficking and Degradation of the Chemokine Receptor 4 at the Sorting Endosome
Berlin, I.
The Journal of Biological Chemistry, 285, 37895?37908-37895?37908 (2006)
S Naviglio et al.
The EMBO journal, 17(12), 3241-3250 (1998-06-17)
The ubiquitin pathway has been implicated in the regulation of the abundance of proteins that control cell growth and proliferation. We have identified and characterized a novel human ubiquitin isopeptidase, UBPY, which both as a recombinant protein and upon immunoprecipitation
Emi Mizuno et al.
Molecular biology of the cell, 16(11), 5163-5174 (2005-08-27)
Ligand-activated receptor tyrosine kinases undergo endocytosis and are transported via endosomes to lysosomes for degradation. This "receptor down-regulation" process is crucial to terminate the cell proliferation signals produced by activated receptors. During the process, ubiquitination of the receptors serves as
Sandra Niendorf et al.
Molecular and cellular biology, 27(13), 5029-5039 (2007-04-25)
Posttranslational modification by ubiquitin controls multiple cellular functions and is counteracted by the activities of deubiquitinating enzymes. UBPy (USP8) is a growth-regulated ubiquitin isopeptidase that interacts with the HRS-STAM complex. Using Cre-loxP-mediated gene targeting in mice, we show that lack

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service