All Photos(1)
About This Item
Linear Formula:
C22H15N2O7S2Na
CAS Number:
Molecular Weight:
506.48
EC Number:
UNSPSC Code:
12171500
composition
Dye content, ~75%
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service