Skip to Content
Merck
All Photos(1)

Key Documents

U106

Sigma-Aldrich

(−)-cis-(1S,2R)-U-50488 tartrate

solid

Synonym(s):

(−)-(1S,2R)-cis-3,4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26Cl2N2O · C4H6O6
CAS Number:
Molecular Weight:
519.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

color

white

solubility

ethanol: soluble

storage temp.

2-8°C

SMILES string

OC(C(O)C(O)=O)C(O)=O.CN([C@H]1CCCC[C@H]1N2CCCC2)C(=O)Cc3ccc(Cl)c(Cl)c3

InChI

1S/C19H26Cl2N2O.C4H6O6/c1-22(19(24)13-14-8-9-15(20)16(21)12-14)17-6-2-3-7-18(17)23-10-4-5-11-23;5-1(3(7)8)2(6)4(9)10/h8-9,12,17-18H,2-7,10-11,13H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t17-,18+;/m0./s1

InChI key

DMBKHRMAGYYBJM-CJRXIRLBSA-N

Gene Information

human ... OPRS1(10280)

Biochem/physiol Actions

Potent σ receptor ligand.

Preparation Note

(-)-cis-(1S,2R)-U-50488 tartrate is soluble in ethanol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Spasov et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(8), 45-47 (2012-01-12)
A new model is proposed for in vitro testing the efficiency of new chemical substances with kappa-opioid receptor agonist ligand properties.
Amber D Shaffer et al.
Neuroscience letters, 534, 150-154 (2012-12-04)
Previous research has suggested that early-in-life (EIL) exposure to bladder inflammation impairs the function of endogenous opioid inhibitory system(s) and may contribute to the development of chronic bladder pain. This study examined how acute adult and/or prior EIL exposure to
K Rasakham et al.
Neuroscience, 223, 447-456 (2012-08-07)
Recently there has been a widespread interest in the development of kappa opioid receptor (KOPR) ligands for treatment of pain, depression and anxiety, and prevention of stress-induced drug relapse. However, most of these preclinical studies have been conducted using male
Q-Schick Auh et al.
Neuroscience letters, 524(2), 111-115 (2012-07-24)
Activation of peripheral κ opioid receptors (KOR) effectively relieves pain and hyperalgesia in preclinical and clinical models of pain. Although centrally located KOR activation results in sexually dimorphic effects, it is unclear whether peripheral KOR also produces sex dependent effects
Yu-Wei Chen et al.
Alcohol (Fayetteville, N.Y.), 47(1), 31-38 (2012-12-04)
The opioid system is known to enhance motivated behaviors, including ethanol drinking and food ingestion, by acting in various reward-related brain regions, such as the nucleus accumbens, ventral tegmental area and medial hypothalamus. There is indirect evidence, however, suggesting that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service