Key Documents
TMO3665-1000
Nalgene® economy graduated cylinder
PMP, volume 1000 mL, case of 6 ea
Synonym(s):
PMP graduated cylinder, PMP graduated flask
Select a Size
6 EACH
€217.00
€217.00
Please contact Customer Service for Availability
Select a Size
Change View
6 EACH
€217.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€217.00
Please contact Customer Service for Availability
material
PMP
packaging
case of 6 ea
manufacturer/tradename
Nalgene 3665-1000
capacity
1000 mL
volume
1000 mL
General description
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)