Skip to Content
Merck
All Photos(1)

Documents

TMO312114-0006

Nalgene® wide-mouth square bottle

high-density polyethylene, capacity 175 mL, natural, case of 250 ea

Synonym(s):

clear plastic bottle, translucent bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

high-density polyethylene
square
wide-mouth

packaging

case of 250 ea

manufacturer/tradename

Nalgene 312114-0006

capacity

175 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

Easily fill and pour with Thermo Scientific Nalgene Square Wide-Mouth HDPE Bottles with Closure, featuring molded-in graduations for easy measuring and durable, uniform walls that are resistant to splitting or puncturing. These translucent bottles provide visible liquid levels and come in a space-saving square design.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service