Skip to Content
Merck
All Photos(4)

Key Documents

T9784

Sigma-Aldrich

Tricine

BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)

Synonym(s):

N-[Tris(hydroxymethyl)methyl]glycine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€66.50
100 G
€204.00
250 G
€457.00
1 KG
€1,540.00

€66.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€66.50
100 G
€204.00
250 G
€457.00
1 KG
€1,540.00

About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
1937804
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

€66.50


Please contact Customer Service for Availability

Request a Bulk Order

product line

BioXtra

Assay

≥99% (titration)

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

pH

4.0-6.0 (1 M in H2O)

useful pH range

7.4-8.8

pKa (25 °C)

8.1

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.1%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

absorption

≤0.05 at 280 in H2O at 1 M
≤0.1 at 260 in H2O at 1 M

application(s)

diagnostic assay manufacturing

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The name Tricine is derived from "tris" and "glycine". Tricine and other good buffers serve as efficient scavengers of hydroxyl radicals in a study of radiation-induced membrane damage. Tricine is widely used as an electrophoresis buffer and for the resuspension of cell pellets. Due to its lower negative charge compared to glycine, it migrates faster. It has a high ionic strength which leads to more ion movement and less protein movement. This aids the separation of low molecular weight proteins in lower percent acrylamide gels. Tricine is also used as a trailing ion that permits the resolution of small proteins at lower acrylamide concentrations than in glycine-sodium dodecyl-sulfate polyacrylamide gel electrophoresis (SDS-PAGE) systems.[1]

Application

Buffer component for separation of low molecular weight peptides.
Tricine has been used:
  • in a polymerase chain reaction (PCR) buffer used in the single-cell collection[2]
  • as a component of lactotroph feeding medium for culturing lactotrophs[3]
  • to prepare luciferase assay reagent for luciferase activity assay in firefly Photonis pyralis and as a component in Triton/glycylglycine lysis buffer[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joke Mertens et al.
Bio-protocol, 9(13), e3283-e3283 (2019-07-05)
Detecting heteroplasmies in the mitochondrial DNA (mtDNA) has been a challenge for many years. In the past, Sanger sequencing was the main option to perform this analysis, however, this method could not detect low frequency heteroplasmies. Massive Parallel Sequencing (MPS)
Encyclopedia of Dairy Sciences (2011)
Analysis of gene promoter regulation by tumor suppressor genes.
Kumaravel Somasundaram et al.
Methods in molecular biology (Clifton, N.J.), 223, 101-116 (2003-06-05)
Boštjan Rituper et al.
Nature protocols, 8(6), 1169-1183 (2013-05-25)
In order to understand exocytosis and endocytosis, it is necessary to study these processes directly. An elegant way to do this is by measuring plasma membrane capacitance (C(m)), a parameter proportional to cell surface area, the fluctuations of which are
Arpita Gantayet et al.
Biofouling, 29(1), 77-85 (2012-12-06)
The freshwater zebra mussel (Dreissena polymorpha) is a notorious biofouling organism. It adheres to a variety of substrata underwater by means of a proteinaceous structure called the byssus, which consists of a number of threads with adhesive plaques at the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service