Skip to Content
Merck
All Photos(1)

Key Documents

T8910

Sigma-Aldrich

β-(2-Thienyl)-D-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO2S
CAS Number:
Molecular Weight:
171.22
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

technique(s)

ligand binding assay: suitable

SMILES string

N[C@H](Cc1cccs1)C(O)=O

InChI

1S/C7H9NO2S/c8-6(7(9)10)4-5-2-1-3-11-5/h1-3,6H,4,8H2,(H,9,10)/t6-/m1/s1

InChI key

WTOFYLAWDLQMBZ-ZCFIWIBFSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Wapnir et al.
The Biochemical journal, 177(1), 347-352 (1979-01-01)
The inhibitory properties of beta-2-thienyl-dl-alanine on rat phenylalanine hydroxylase from crude liver and kidney homogenates were assessed in vitro and in vivo, as well as its effects on the intestinal transport of phenylalanine, by using a perfusion procedure in vivo.
X Liu et al.
Biopolymers, 33(8), 1237-1247 (1993-08-01)
Bradykinin (BK) is a peptide hormone with sequence Arg1-Pro2-Pro3-Gly4-Phe5-Ser6-Pro7-Phe8-Arg9 and has been implicated in a multitude of pathophysiological processes such as the ability to lower systemic blood pressure and stimulate pain. BK analogues having bulky, beta-branched D-aliphatic residues at position
Uptake and incorporation of amino acids by suspension cultured mammalian cells: a comparative study involving eleven naturally-occurring and four analogue amino acids.
D N Wheatley et al.
Cytobios, 30(118), 101-126 (1981-01-01)
"Physiological phenylketonuria": a biochemical defect caused by delayed maturation of the phenylalanine hydroxylation pathway and by competition with the phenylalanine biosynthetic pathway.
K J Brown
Medical hypotheses, 6(2), 215-224 (1980-02-01)
D P Bedard et al.
Journal of bacteriology, 141(1), 100-105 (1980-01-01)
When treated with the amino acid analog beta-2-DL-thienylalanine, cells of the yeast Saccharomyces cerevisiae accumulated in the G1 portion of the cell cycle at the "start" event. This G1 arrest was accompanied by a rapid decrease in the rate of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service