Skip to Content
Merck
All Photos(1)

Documents

T821020

Sigma-Aldrich

dT Polystyrene column

configured for ABI 3900, 0.2μmol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥97.0% (Nucleoside purity, reversed phase HPLC)

compatibility

configured for ABI 3900

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service