Skip to Content
Merck
All Photos(1)

Key Documents

T6943

Sigma-Aldrich

Tyrphostin AG 112

≥90% (NMR)

Synonym(s):

3-Amino-2,4-dicyano-5-(4′-hydroxyphenyl)penta-2,4-dienonitrile, Tyrphostin A48

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N4O
Molecular Weight:
236.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90% (NMR)

potency

125 nM IC50

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

NC(\C(=C/c1ccc(O)cc1)C#N)=C(\C#N)C#N

Biochem/physiol Actions

Protein tyrosine kinase inhibitor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trine Reistad et al.
Toxicological sciences : an official journal of the Society of Toxicology, 87(1), 57-65 (2005-06-17)
Polybrominated diphenyl ethers (PBDEs) are widely used brominated flame retardants (BFRs), which have become ubiquitous in the environment. This study investigates the effects of the pentabrominated diphenyl ether mixture, DE-71, on human neutrophil granulocytes in vitro. DE-71 enhanced production of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service