Skip to Content
Merck
All Photos(1)

Documents

T6514

Sigma-Aldrich

Oleandomycin triacetate

Synonym(s):

Troleandomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H67NO15
CAS Number:
Molecular Weight:
813.97
Beilstein:
1418238
EC Number:
MDL number:
UNSPSC Code:
51101500

Mode of action

enzyme | inhibits
protein synthesis | interferes

SMILES string

CO[C@H]1C[C@@H](O[C@@H](C)[C@@H]1OC(C)=O)O[C@H]2[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3OC(C)=O)N(C)C)[C@@H](C)C[C@@]4(CO4)C(=O)[C@H](C)[C@@H](OC(C)=O)[C@@H](C)[C@@H](C)OC(=O)[C@@H]2C

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo H Bae et al.
The Journal of pharmacy and pharmacology, 61(12), 1637-1642 (2009-12-05)
It has been reported that hepatic cytochrome P450 (CYP)2C9 and CYP3A4 are responsible for the metabolism of sildenafil and formation of its metabolite, N-desmethylsildenafil, in humans. However, in-vivo studies in rats have not been reported. Sildenafil (20 mg/kg) was administered
Shobana Ganesan et al.
Toxicology and applied pharmacology, 241(1), 14-22 (2009-07-21)
Primaquine, an 8-aminoquinoline, is the drug of choice for radical cure of relapsing malaria. Use of primaquine is limited due to its hemotoxicity, particularly in populations with glucose-6-phosphate dehydrogenase deficiency [G6PD(-)]. Biotransformation appears to be central to the anti-infective and
Daniel R Albaugh et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(7), 1336-1344 (2012-04-12)
The current studies assessed the utility of freshly plated hepatocytes, cryopreserved plated hepatocytes, and cryopreserved plated HepaRG cells for the estimation of inactivation parameters k(inact) and K(I) for CYP3A. This was achieved using a subset of CYP3A time-dependent inhibitors (fluoxetine
Güliz Gürel et al.
Antimicrobial agents and chemotherapy, 53(12), 5010-5014 (2009-09-10)
Structures have been obtained for the complexes that triacetyloleandomycin and mycalamide A form with the large ribosomal subunit of Haloarcula marismortui. Triacetyloleandomycin binds in the nascent peptide tunnel and inhibits the activity of ribosomes by blocking the growth of the
Y H Choi et al.
British journal of pharmacology, 149(4), 424-430 (2006-08-31)
The types of hepatic microsomal cytochrome P450 (CYP) isozymes responsible for the metabolism of metformin in humans and rats have not been published to date. Therefore, a series of experiments using various inducers and inhibitors of CYP isozymes was conducted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service