This product is soluble in water at approximately 2 mg/ml.
T5771
Testosterone 3-(O-carboxymethyl)oxime: BSA - fluorescein isothiocyanate conjugate
Select a Size
About This Item
Recommended Products
Assay
≥98.00% (TLC)
Quality Level
form
powder
drug control
regulated under CDSA - not available from Sigma-Aldrich Canada
extent of labeling
~3 mol FITC per mol BSA
~10 mol steroid per mol BSA
technique(s)
flow cytometry: suitable
solubility
water: 1.90-2.10 mg/mL, clear to faintly hazy, orange
shipped in
ambient
storage temp.
2-8°C
Application
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How will you dissolve this product (T5771)?
1 answer-
Helpful?
-
-
How can I dissolve the products that arrived in solid form?
1 answer-
The spec sheet indicates that the product is tested for solubility at 2mg/ml in water. For Testosterone 3-(O-carboxymethyl)oxime: BSA, solubility is tested in Phosphate buffer pH 7.5. Additionally, without the albumin conjugate, Testosterone 3-(O-carboxymethyl)oxime is soluble in ethanol at 50mg/ml.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service