T5395
TAPSO sodium salt
Synonym(s):
3-(N-tris[Hydroxymethyl]methylamino)-2-hydroxypropanesulfonic acid
About This Item
Recommended Products
Assay
≥99% H2SO4 basis (titration)
form
powder or crystals
useful pH range
7.0-8.2
pKa (25 °C)
7.6
solubility
water: 0.333 g/mL, clear to slightly hazy, colorless
application(s)
diagnostic assay manufacturing
SMILES string
[Na+].OCC(CO)(CO)NCC(O)CS([O-])(=O)=O
InChI
1S/C7H17NO7S.Na/c9-3-7(4-10,5-11)8-1-6(12)2-16(13,14)15;/h6,8-12H,1-5H2,(H,13,14,15);/q;+1/p-1
InChI key
XRADYQRMWVBZEH-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service