Skip to Content
Merck
All Photos(1)

Key Documents

T5050

Sigma-Aldrich

Transforming Growth Factor-β1 from porcine platelets

≥97% (SDS-PAGE), recombinant, powder, suitable for cell culture

Synonym(s):

TGF-β1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Transforming Growth Factor-β1 from porcine platelets, TGF-β1, powder, suitable for cell culture

biological source

Porcine (Platelets)

Quality Level

Assay

≥97% (SDS-PAGE)

form

powder

potency

0.01-0.1 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

protein 25 kDa

packaging

pkg of 1 μg
pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

The TGFB (transforming growth factor-β) gene is mapped to pig (Sus scrofa) chromosome between 6q11-q21. In mammals, TGFβ is present in three isoforms (TGFβ1, TGFβ2 and TGFβ3), of which TGFβ1 is predominant.

Application

Transforming Growth Factor-β1 from porcine platelets has been used in chondrogenic differentiation. It has also been used in immobilization process.

Biochem/physiol Actions

Transforming growth factor-β1 (TGF-β) is a multifunctional polypeptide growth factor. TGF-β mediates cell proliferation, differentiation and migration. Important cellular functions such as inflammation, extracellular matrix synthesis, epithelial restoration and developmental processes are also controlled by TGF-β. TGF-β controls the regulation of enamel mineralization in porcine. TGF-β is associated with the development of idiopathic pulmonary fibrosis (a chronic lung disease). TGF-β promotes epithelial-mesenchymal transition (EMT) and contributes to proliferation and metastasis of cancer. The TGF-β induced EMT is observed in kidney fibrosis. TGF-β is stimulates regeneration of intestinal tissue.
Transforming growth factor-β1 (TGF-β1) is produced by many cell types, but is reported to be most concentrated in mammalian platelets, where it is present at approximately four times the level of TGF-β2.

Physical form

Lyophilized from a 0.2 μm filtered solution in 35% acetonitrile and 0.1% trifluoroacetic acid containing 0.05 mg bovine serum albumin per μg cytokine.

Analysis Note

The biological activity is measured by its ability to inhibit the IL-4-dependent proliferation of mouse HT-2 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TGF-β1 autocrine signalling and enamel matrix components
Kinoshita SK, et al.
Scientific Reports (2016)
Focal adhesion kinase signaling determines the fate of lung epithelial cells in response to TGF-β
Ding Q, et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 312(6), L926-L935 (2017)
Isolation, culture and chondrogenic differentiation of canine
adipose tissue- and bone marrow-derived mesenchymal stem
cells?a comparative study
Reich CM, et al.
Veterinary Research Communications, 36, 139-148 (2012)
Effect of Conceptus on Transforming Growth Factor (TGF) β1 mRNA Expression and Protein Concentration in the Porcine Endometrium?In Vivo and In Vitro Studies
Blitek A, et al.
Journal of Reproduction and Development, 59(6), 512-519 (2013)
Developmental Changes of TGF-β1 and Smads Signaling Pathway in Intestinal Adaption of Weaned Pigs
Xiao K, et al.
PLoS ONE, 9(8) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service