Skip to Content
Merck
All Photos(1)

Key Documents

T4924

Sigma-Aldrich

Theophylline-[8-3H]

aqueous ethanol solution (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H8O2N4
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

aqueous ethanol solution (1:1)

extent of labeling

15-30 Ci/mmol

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]c1nc2N(C)C(=O)N(C)C(=O)c2[nH]1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoto Fujii et al.
The Journal of physiology, 593(11), 2515-2525 (2015-03-27)
In humans in vivo, the mechanisms behind ATP-mediated cutaneous vasodilatation along with whether and how ATP increases sweating remains uncertain. Recent work has implicated nitric oxide synthase (NOS), cyclooxygenase (COX) and/or adenosine in the modulation of cutaneous vasodilatation and sweat
Rania O Salama et al.
Drug delivery and translational research, 4(3), 256-267 (2015-03-20)
A novel approach to concurrently deliver oral and inhaled drugs as a single formulation is presented. A triple therapy containing theophylline (THEO; orally delivered) with budesonide (BUD) and terbutaline (TERB; as single and co-spray-dried inhaled powders) was prepared as an
Lauriane Goldwirt et al.
Cancer chemotherapy and pharmacology, 74(1), 185-193 (2014-05-29)
Glioblastoma (GBM), the most common primary brain tumor in adults, is usually rapidly fatal with median survival duration of only 15 months and a 3-year survival rate of <7 %. Temozolomide (TMZ) is the only anticancer drug that has improved
Diego Caccavo et al.
Molecular pharmaceutics, 12(2), 474-483 (2014-12-17)
In this work the behavior of hydrogel-based matrices, the most widespread systems for oral controlled release of pharmaceuticals, has been mathematically described. In addition, the calculations of the model have been validated against a rich set of experimental data obtained
Laura López-Cruz et al.
Behavioural brain research, 270, 213-222 (2014-05-27)
Caffeine and theophylline are methylxanthines that are broadly consumed, sometimes at high doses, and act as minor psychostimulants. Both are nonselective adenosine antagonists for A1 and A2A receptors, which are colocalized with dopamine D1 and D2 receptors in striatal areas.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service