Skip to Content
Merck
All Photos(1)

Documents

T4577

Sigma-Aldrich

Terodiline hydrochloride

≥98% (HPLC), solid

Synonym(s):

Bicor, N-(1,1-Dimethylethyl)-alpha-methyl-gamma-phenylbenzenepropamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27N·HCl
CAS Number:
Molecular Weight:
317.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

Cl.CC(CC(c1ccccc1)c2ccccc2)NC(C)(C)C

InChI

1S/C20H27N.ClH/c1-16(21-20(2,3)4)15-19(17-11-7-5-8-12-17)18-13-9-6-10-14-18;/h5-14,16,19,21H,15H2,1-4H3;1H

InChI key

RNGHAJVBYQPLAZ-UHFFFAOYSA-N

Biochem/physiol Actions

Terodiline hydrochloride is a non-selective calcium channel antagonist with anticholinergic and vasodilatory activity.
Terodiline is known to increase corrected QT interval (QTc) and decrease resting heart rate in elderly patients. Furthermore, terodiline hydrochloride has been linked to cardiac arrhythmias1.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Terodiline hydrochloride is soluble in DMSO at a concentration that is greater than 20 mg/ml.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S E Jones et al.
British journal of pharmacology, 125(6), 1138-1143 (1998-12-24)
Prolongation of the QT interval and malignant ventricular arrhythmia have been observed in patients administered terodiline for urinary incontinence. Since this adverse reaction might be caused by inhibition of delayed-rectifier K+ current (IK), we investigated whether clinically relevant (< or
ChangJiang Xu et al.
PloS one, 7(8), e41694-e41694 (2012-08-24)
The investigation of associations between rare genetic variants and diseases or phenotypes has two goals. Firstly, the identification of which genes or genomic regions are associated, and secondly, discrimination of associated variants from background noise within each region. Over the
Leah R Gerber et al.
PloS one, 5(8), e12230-e12230 (2010-09-03)
In polygynous mating systems, males often increase their fecundity via aggressive defense of mates and/or resources necessary for successful mating. Here we show that both male and female reproductive behavior during the breeding season (June-August) affect female fecundity, a vital
Ruth L Martin et al.
Journal of cardiovascular pharmacology, 48(5), 199-206 (2006-11-18)
Terodiline and tolterodine are drugs used to treat urinary incontinence. Terodiline was removed from the market in 1991 for proarrhythmia, whereas tolterodine has a generally benign clinical cardiac profile. To assess differences in the electrophysiologic actions of these drugs, we
K Hartigan-Go et al.
Clinical pharmacology and therapeutics, 60(1), 89-98 (1996-07-01)
To study the cardiovascular and electrocardiographic (ECG) effects of the R(+)- and S(-)- enantiomers of terodiline. The racemic drug was previously used to treat detrusor instability but was withdrawn after it caused serious ventricular arrhythmias associated with prolongation of the

Articles

We offers many products related to calcium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service