Skip to Content
Merck
All Photos(1)

Documents

T3013

Sigma-Aldrich

D-allo-Threonine

Synonym(s):

(2R,3R)-2-Amino-3-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721644
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

mp

276 °C (dec.) (lit.)

SMILES string

C[C@@H](O)[C@@H](N)C(O)=O

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service