Skip to Content
Merck
All Photos(1)

Key Documents

T25

Sigma-Aldrich

5′-Nucleotides Sodium Salts

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.51

shipped in

dry ice

storage temp.

−20°C

Application

5′′-TriNucleotides are used for the synthesis of ribonucleic acids with RNA polymerases.

Other Notes

4 Nucleoside Triphosphates in quantities as indicated
A 2383   ATP, Disodium, 100 mg
C 9274   CTP, Sodium, 25 mg
G 8877   GTP, Sodium, 25 mg
U 6625   UTP, Sodium, 25 mg

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyung Sub Park et al.
Stem cells international, 2018, 4162075-4162075 (2018-07-17)
The use of nanomaterials for biomedical applications has become a promising field in regenerative medicine. Self-assembling peptides (SAPs) have been proposed as a good candidate because they are able to self-assemble into stable hydrogels and interact with cells or molecules
Micaela Cunha et al.
Radiation and environmental biophysics, 55(1), 31-40 (2015-12-29)
The biological phenomena observed at low doses of ionizing radiation (adaptive response, bystander effects, genomic instability, etc.) are still not well understood. While at high irradiation doses, cellular death may be directly linked to DNA damage, at low doses, other
Narumi Uno et al.
Scientific reports, 7(1), 12739-12739 (2017-10-08)
Chromosome engineering techniques including gene insertion, telomere-associated truncation and microcell-mediated chromosome transfer (MMCT) are powerful tools for generation of humanised model animal, containing megabase-sized genomic fragments. However, these techniques require two cell lines: homologous recombination (HR)-proficient DT40 cells for chromosome
Silvia Mrvová et al.
Molecular genetics and genomics : MGG, 293(1), 167-186 (2017-09-25)
Alternative polyadenylation is an important and pervasive mechanism that generates heterogeneous 3'-termini of mRNA and is considered an important regulator of gene expression. We performed bioinformatics analyses of ESTs and the 3'-UTRs of the main transcript splice variants of the
David Alan Schneider
Gene, 493(2), 176-184 (2011-09-07)
Synthesis of the translation apparatus is a central activity in growing and/or proliferating cells. Because of its fundamental importance and direct connection to cell proliferation, ribosome synthesis has been a focus of ongoing research for several decades. As a consequence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service