Skip to Content
Merck
All Photos(1)

Key Documents

T211850

Sigma-Aldrich

DMT-2′O-MOE-rMeU Phosphoramidite

Synonym(s):

5′-Dimethoxytrityl-3′-[(2-cyanoethyl)- (N,N-diisopropyl)]-phosphoramidite- 2′-O-(2-methoxyethyl)-5-methyluridine

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€858.00

€858.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€858.00

About This Item

Empirical Formula (Hill Notation):
C43H55N4O10P
CAS Number:
Molecular Weight:
818.89
UNSPSC Code:
41116105
NACRES:
NA.51

€858.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98.0% (reversed phase HPLC)
≥99% (31P-NMR)

Quality Level

λ

conforms (UV/VIS Identity)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′O-MOE-rMeU Phosphoramidite is a phosphoramidite that belongs to the family of amides of trivalent phosphorus acid H3PO3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service