Skip to Content
Merck
All Photos(1)

Key Documents

T1955

Sigma-Aldrich

TOPK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ14385, Nori-3, PBK, SPK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

5-7.5 nmol/min·mg

mol wt

~68 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PBK(55872)

Biochem/physiol Actions

TOPK is a MAPK kinase that phosphorylates p38 MAPK and is activated in a cell-cycle-dependent manner in neuronal progenitor cells in vitro. Expression of TOPK is detected in male germ line progenitor cells, activated T-cells, and a variety of lymphomas and leukemias. In vitro studies have shown that activated TOPK phosphorylated p38MAPK but not JNK or ERK. TOPK activation requires phosphorylation by both the M-phase CDK1/CyclinB kinase complex and another unknown kinase, possibly RafC or RafA. TOPK may play an important role in linking extracellular signals to an intracellular state, possibly allowing extracellular influence on the cell-cycle-related processes of proliferation or differentiation.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suguru Matsumoto et al.
Biochemical and biophysical research communications, 325(3), 997-1004 (2004-11-16)
A MAPKK-like protein kinase TOPK expresses in a wide range of proliferating cells and tissues such as cancer cells and testis. However, details of this kinase are still uncovered. We investigated the intracellular distribution of TOPK and its association with
M Simons-Evelyn et al.
Blood cells, molecules & diseases, 27(5), 825-829 (2002-01-11)
PBK/TOPK is a recently cloned serine/threonine kinase which is phosphorylated during mitosis. Earlier work indicated that this kinase is upregulated in a Burkitt's lymphoma cell line (GA-10). To determine whether PBK/TOPK is upregulated in other mitotically active neoplastic cell lines

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service