Skip to Content
Merck
All Photos(1)

Key Documents

T1647

Supelco

Trifluoroacetic acid

≥99%, liquid

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12190000
Pricing and availability is not currently available.

Assay

≥99%

form

liquid

density

1.49 g/mL at 25 °C (lit.)

Application

TFA can be used as an ion-pairing reagent for reverse-phase chromatography.

Packaging

Flame-sealed ampules contain 1 ml. Other sizes are in screw-cap bottles.

Preparation Note

Distilled from chromium(VI) oxide under nitrogen, treated with aluminum oxide, then redistilled from dithiothreitol under nitrogen.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mangesh R Bhide et al.
BMC research notes, 2, 134-134 (2009-07-17)
Borreliae employ multiple immune evasive strategies such as binding to complement regulatory proteins [factor H (fH) and factor H like-1 (FHL1)], differential regulation of surface membrane proteins, antigenic variation, and binding of plasminogen/plasmin and matrix metalloproteinases. As a complement regulatory
Miklos Guttman et al.
Proteomics, 9(22), 5016-5028 (2009-09-23)
The low density lipoprotein receptor-related protein 1 (LRP1) mediates internalization of a large number of proteins and protein-lipid complexes and is widely implicated in Alzheimer's disease. The cytoplasmic domain of LRP1 (LRP1-CT) can be phosphorylated by activated protein-tyrosine kinases at
Younggyu Kim et al.
Nucleic acids research, 36(19), 6143-6154 (2008-10-01)
We have developed an approach that enables nonradioactive, ultrasensitive (attamole sensitivity) site-specific protein-protein photocrosslinking, and we have applied the approach to the analysis of interactions of alpha-helix 2 (H2) of human TATA-element binding protein (TBP) with general transcription factor TFIIA
Anna Steinle et al.
Applied and environmental microbiology, 75(11), 3437-3446 (2009-04-07)
Cyanophycin (multi-l-arginyl-poly-l-aspartic acid; also known as cyanophycin grana peptide [CGP]) is a putative precursor for numerous biodegradable technically used chemicals. Therefore, the biosynthesis and production of the polymer in recombinant organisms is of special interest. The synthesis of cyanophycin derivatives
Caroline Le Maréchal et al.
Veterinary research, 42, 35-35 (2011-02-18)
Staphylococcus aureus is a major cause of mastitis in ruminants. In ewe mastitis, symptoms range from subclinical to gangrenous mastitis. S. aureus factors or host-factors contributing to the different outcomes are not completely elucidated. In this study, experimental mastitis was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service