Skip to Content
Merck
All Photos(1)

Key Documents

T0267

Sigma-Aldrich

3,4,5,6-Tetrahydrophthalic anhydride

≥95%

Synonym(s):

Cyclohexene-1,2-dicarboxylic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
Beilstein:
136335
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

mp

69-73 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1OC(=O)C2=C1CCCC2

InChI

1S/C8H8O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H2

InChI key

HMMBJOWWRLZEMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subat Turdi et al.
Toxicology letters, 178(3), 197-201 (2008-05-10)
Doxorubicin is an effective anthracycline used for cancer therapy. However, the clinical application of doxorubicin has been largely limited by its irreversible cardiotoxicity, which is mainly induced by the primary amine group. In this study, we structurally modified doxorubicin by
J B Swaney
Journal of lipid research, 24(3), 245-252 (1983-03-01)
Trypsin digestion of human high density lipoprotein (d 1.125-1.21 g/ml) on which the lysine residues have been masked with the reversible blocking group, 2,3,4,5-tetrahydrophthallic anhydride (THPA), was found to result in the fragmentation of the apoA-I component, but not the
L I O'Connell et al.
Archives of biochemistry and biophysics, 263(2), 315-322 (1988-06-01)
Modification of glutamate dehydrogenase with 3,4,5,6-tetrahydrophthalic anhydride at pH 8.0 results in the progressive loss of enzymatic activity and a concomitant increase in the negative charge of the protein. Although the rate of inactivation at room temperature is too rapid
A Ikai et al.
The Journal of biological chemistry, 265(14), 8280-8284 (1990-05-15)
As a model for the molecular structure of proteins belonging to the alpha 2-macroglobulin family, ovomacroglobulin of reptilian origin was studied by electron microscopy in the original tetrameric form as well as in the dissociated forms into half- and quarter
[Data providing a basis for the MPEL of tetrahydrophthalic anhydride, tetrahydrophthalamide and hydroxymethyltetrahydrophthalimide in the air of the work environment].
A I Gurova et al.
Gigiena truda i professional'nye zabolevaniia, (12)(12), 37-38 (1985-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service