Skip to Content
Merck
All Photos(1)

Key Documents

SRP8015

Sigma-Aldrich

Progranulin mouse

>95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized

Synonym(s):

GPx-1, PEPI, Proepithelin

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€297.00

€297.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€297.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€297.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Progranulin mouse, recombinant, expressed in HEK 293 cells, untagged, >95% (SDS-PAGE)

biological source

mouse

recombinant

expressed in HEK 293 cells

conjugate

untagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~65 kDa by SDS-PAGE

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Grn(14824)

General description

Progranulin (PGRN) is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration (FTLD). In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer′s Disease (AD) and potentially for MCI (Mild Cognitive Impairment). Additionally, PGRN is described as a new ligand of TNF receptors and a potential therapeutic against inflammatory disease like arthritis.

Physical form

Lyophilized from 0.2 μm-filtered solution in PBS, pH 7.2.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

Signal peptide and mouse progranulin (aa 1-589) is untagged. Reflects the native sequence with no additional aa.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenz Thurner et al.
Digestive diseases and sciences, 59(8), 1733-1742 (2014-03-05)
Recently, we identified neutralizing autoantibodies against progranulin (PGRN) in a wide spectrum of rheumatic diseases including cases with enteropathic spondylarthritis. PGRN is a secreted protein with strong anti-inflammatory effects, believed to be mediated by the direct inhibition of TNF receptors
Masato Kanazawa et al.
Brain : a journal of neurology, 138(Pt 7), 1932-1948 (2015-04-04)
In the central nervous system, progranulin, a glycoprotein growth factor, plays a crucial role in maintaining physiological functions, and progranulin gene mutations cause TAR DNA-binding protein-43-positive frontotemporal lobar degeneration. Although several studies have reported that progranulin plays a protective role
Michael J Smout et al.
PLoS pathogens, 11(10), e1005209-e1005209 (2015-10-21)
Infection with the human liver fluke Opisthorchis viverrini induces cancer of the bile ducts, cholangiocarcinoma (CCA). Injury from feeding activities of this parasite within the human biliary tree causes extensive lesions, wounds that undergo protracted cycles of healing, and re-injury

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service