Skip to Content
Merck
All Photos(1)

Key Documents

SRP8007

Sigma-Aldrich

DLK2 (human): FC (human)

recombinant, expressed in HEK 293 cells, >90% (SDS-PAGE)

Synonym(s):

C1QTNF3, Epidermal growth factor-like protein 9, Protein delta homolog 2 (DLK-2)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€429.00

€429.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€429.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€429.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in HEK 293 cells

Assay

>90% (SDS-PAGE)

form

liquid

mol wt

~70 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

≥0.2 mg/mL

impurities

<0.1 EU/μg endotoxin, tested

color

clear

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DLK2(65989)

Related Categories

General description

DLK2 regulates adipogenesis.

Physical form

Solution in PBS.

Other Notes

Signal peptide and the extracellular domain of human DLK2 (aa 1-306) are fused at the C-terminus to the Fc portion of human IgG.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service