Skip to Content
Merck
All Photos(1)

Key Documents

SRP6432

Sigma-Aldrich

CD58/LFA-3 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Ag3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 22.3 kDa
observed mol wt 30-45 kDa (DTT-reduced. Protein migrates due to glycosylation. Phe 29 is the predicted N-terminal.)

packaging

pkg of 10 μg
pkg of 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD58(965)

General description

Cluster of differentiation 58 (CD58), also known as lymphocyte function-associated antigen 3 (LFA-3), is a cell adhesion molecule expressed on antigen presenting cells (APC), particularly macrophages.[1] The gene encoding this protein is localized on human chromosome 1p13.1.[2]
CD58 /LFA3 is ligand of the T-lymphocyte CD2 glycoprotein, which binds to CD2 (LFA-2) on T cells and is important in strengthening the adhesion between the T cells and Professional Antigen Presenting Cells. In addition, the LFA-3/CD2 interaction may prime response by both the CD2+ and LFA-3+ cells. Polymorphisms in the CD58 gene are associated with increased risk for multiple sclerosis.

Biochem/physiol Actions

Cluster of differentiation 58 (CD58)/lymphocyte function-associated antigen 3 (LFA-3) is a ligand of the T-lymphocyte cluster of differentiation 2 (CD2) glycoprotein, which binds to CD2 (LFA-2) on T cells.[1] It is important in strengthening the adhesion between the T cells and professional antigen presenting cells. In addition, the LFA-3/CD2 interaction may prime response by both the CD2+ and LFA-3+ cells. Polymorphisms in the CD58 gene are associated with increased risk for multiple sclerosis.[3]

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular adhesion molecule-1 modulates β-chemokines and directly costimulates T cells in vivo.
Kim J J., et al.
The Journal of Clinical Investigation, 103(6), 869-877 (1999)
Genetic variants at CD28, PRDM1 and CD2/CD58 are associated with rheumatoid arthritis risk.
Raychaudhuri S, et al.
Nature Genetics, 41(12), 1313-1313 (2009)
CD58 polymorphisms associated with the risk of neuromyelitis optica in a Korean population.
Kim J Y, et al.
BMC Neurology, 14(1), 57-57 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service