Skip to Content
Merck
All Photos(1)

Key Documents

SRP6263

Sigma-Aldrich

PLGF/PIGF2/PGF human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

PGFL, SHGC-10760

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 20.2 kDa
observed mol wt 28-33 kDa (DTT-reduced. Protein migrates due to glycosylation. Arg 29 is the predicted N-terminus.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PLGF(5228)

General description

Placental growth factor (PLGF) exists in two isoforms namely PLGF1 and PLGF2. This protein is a homodimeric glycoprotein with molecular weight of 46–50kDa and is a member of the VEGF (vascular endothelial growth factor) subfamily. PLGF is encoded by the gene mapped to human chromosome 14q24-q31 regions. PLGF is expressed mainly in the placental trophoblast during pregnancy and is also present in various other tissues such as the villous trophoblast.

Biochem/physiol Actions

PLGF might enhance the production of peripheral blood mononuclear cell-vascular endothelial growth factor (PBMC-VEGF). Consequently, PlGF might facilitate VEGF production in the inflammatory joint. Soluble fms related tyrosine kinase 1 (Flt-1), which is considered to be an anti-angiogenic factor interacts with and inhibit the activity of PLGF. Therefore, PLGF level is lowered and the Flt-1 level is increased in placental tissue of women with pre-eclampsia (PE). PLGF is associated with the pathogenesis of rheumatoid arthritis (RA) and various other arthropathies. PLGF acts as a ligand for VEGFR-1 and is implicated in various physiological functions such as tumor angiogenesis, wound healing and hematopoietic progenitors recruitment.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of antihypertensive therapy with alpha methyldopa on levels of angiogenic factors in pregnancies with hypertensive disorders.
Khalil A
PLoS ONE, 3(7), e2766-e2766 (2008)
Role of placenta growth factor (PIGF) in human extravillous trophoblast proliferation, migration and invasiveness.
Athanassiades A and Lala PK
Placenta, 19(7), 465-473 (1998)
PlGF blockade does not inhibit angiogenesis during primary tumor growth.
Bais C
Cell, 141(1), 166-177 (2010)
Placenta growth factor (PlGF) induces vascular endothelial growth factor (VEGF) secretion from mononuclear cells and is co-expressed with VEGF in synovial fluid.
Bottomley MJ
Clinical and Experimental Immunology, 119(1), 182-188 (2000)
Assignment of vascular endothelial growth factor (VEGF) and placenta growth factor (PLGF) genes to human chromosome 6p12-p21 and 14q24-q31 regions, respectively.
M G Mattei et al.
Genomics, 32(1), 168-169 (1996-02-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service