Skip to Content
Merck
All Photos(1)

Key Documents

SRP6182

Sigma-Aldrich

Human Serum Albumin

≥98% (SDS-PAGE)

Synonym(s):

GIG20, GIG42, HSA, PRO0903, PRO1708, PRO2044, PRO2619, PRO2675, UNQ696/PRO1341, serum albumin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

Pricing and availability is not currently available. Contact Technical Service

biological source

human

Assay

≥98% (SDS-PAGE)

form

liquid

mol wt

66.4 kDa

packaging

pkg of 1 mg

concentration

10 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... ALBU(213)

General description

The gene HSA (human serum albumin) is mapped to human chromosome 4q13.3.[1] The gene encodes a protein of 585 amino acids with a molecular weight of 66.4kDa.[2] HSA is the most abundant protein in the plasma and has many metal-binding sites.[3][4]

Application

HSA (human serum albumin) has been used for the development of automated dye-binding method for serum albumin determination using bromcresol purple.[5] It has also been used to study effect of HSA on DNA transfection by lipoplexes.[6]

Biochem/physiol Actions

HSA (human serum albumin) mainly works as a carrier for various nutrients, metabolites and xenobiotics. It interacts with plasma zinc and regulates uptake of zinc in cells.[4] HSA plays a crucial role in drug pharmacokinetics.[3] It also controls oncotic pressure and volume of the blood.[7] Mutations in the gene are linked with congenital analbuminemia and familial dysalbuminemic hyperthyroxinemia.[1][8]

Physical form

Sterile filtered yellowish liquid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and ligand binding properties of human serum albumin.
Kragh-Hansen U
Danish Medical Bulletin, 37, 57-84 (1990)
New automated dye-binding method for serum albumin determination with bromcresol purple.
Pinnell AE and Northam BE
Clinical Chemistry, 24, 80-86 (1978)
Structural basis of non-steroidal anti-inflammatory drug diclofenac binding to human serum albumin.
Zhang Y
Chemical Biology & Drug Design, 86, 1178-1184 (2015)
An identical missense mutation in the albumin gene results in familial dysalbuminemic hyperthyroxinemia in 8 unrelated families.
Sunthornthepvarakul T
Biochemical and Biophysical Research Communications, 202, 781-787 (1994)
Human serum albumin enhances DNA transfection by lipoplexes and confers resistance to inhibition by serum.
Sim?es S
Biochimica et Biophysica Acta, 1463, 459-469 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service