SRP6065
Hexokinase 1 human
recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonym(s):
Brain form hexokinase, HK1, Hexokinase type I, N/A
Select a Size
€959.00
Select a Size
About This Item
€959.00
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥90% (SDS-PAGE)
form
liquid
potency
7-8 units/mL
mol wt
104.6 kDa
packaging
pkg of 100 μg
storage condition
avoid repeated freeze/thaw cycles
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... HXK1(3098)
General description
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Isozymes of mammalian hexokinase: structure, subcellular localization and metabolic function.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service