Skip to Content
Merck
All Photos(1)

Key Documents

SRP6023

Sigma-Aldrich

Growth Hormone from chicken

recombinant, expressed in E. coli, ≥99% (SDS-PAGE), ≥99% (HPLC)

Synonym(s):

Growth hormone, Growth hormone 1, Somatotropin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

chicken

recombinant

expressed in E. coli

Assay

≥99% (HPLC)
≥99% (SDS-PAGE)

form

lyophilized

mol wt

22.255 kDa

packaging

pkg of 50 μg

impurities

Endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

chicken ... SOMA(378781)

General description

GH is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The gene, along with four other related genes, is located at the growth hormone locus on chromosome 17 where they are interspersed in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. The five genes share a remarkably high degree of sequence identity. Alternative splicing generates additional isoforms of each of the five growth hormones, leading to further diversity and potential for specialization. This particular family member is expressed in the pituitary but not in placental tissue as is the case for the other four genes in the growth hormone locus. Growth Hormone Chicken Recombinant (cGH) produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 191 amino acids with an additional Ala at its N-terminus and having a molecular mass of 22255 Dalton. GH Chicken recombinant is purified by proprietary chromatographic techniques.

Physical form

Lyophilized from 0.3 % NaHCO?, pH 8

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile H?O or 0.4 % NaHCO? adjusted to pH 8-9 to a concentration ? 100 μg/ml and sterile filtered. This solution can then be diluted into other aqueous buffers, preferably in the presence of carrier protein.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service