Skip to Content
Merck
All Photos(1)

Key Documents

SRP5373

Sigma-Aldrich

INCENP, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ31633, MGC111393

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~19 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... INCENP(3619)

General description

INCENP or inner centromere protein antigens 135/155kDa is a constitutively binding centromere proteins and ′passenger,′ or transiently interacting, proteins. The overexpression of a truncation mutant of chicken INCENP and a chimeric CENPB:INCENP protein in mammalian cell culture disrupted both prometaphase chromosome alignment and completion of cytokinesis. INCENP forms a complex with the evolutionarily conserved family of Aurora B kinases which helps coordinate chromosome segregation, spindle behavior, and cytokinesis during mitosis.

Biochem/physiol Actions

As part of the chromosomal passenger complex (CPC), inner centromere protein (INCENP) has a role in assembly and stability of the complex. It also aids in the centromere localization of CPC. INCENP has a role in mitosis and associates with microtubules. The protein has been shown to be upregulated in colorectal cancer cell lines.[1][2][3]

Physical form

Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human INCENP colocalizes with the Aurora-B/AIRK2 kinase on chromosomes and is overexpressed in tumour cells.
Adams RR, et.al
Chromosoma, 110(2), 65-74 (2001)
INCENP binds directly to tubulin and requires dynamic microtubules to target to the cleavage furrow.
Wheatley SP, et.al
Experimental Cell Research, 262(2), 122-127 (2001)
Inherited variants in the inner centromere protein (INCENP) gene of the chromosomal passenger complex contribute to the susceptibility of ER-negative breast cancer.
Kabisch M, et.al
Carcinogenesis, 36(2), 256-271 (2015)
S P Wheatley et al.
Experimental cell research, 262(2), 122-127 (2001-01-05)
Inner centromere protein (INCENP) is a chromosomal passenger protein with an essential role in mitosis. At the metaphase/anaphase transition, some INCENP transfers from the centromeres to the central spindle; the remainder then transfers to the equatorial cortex prior to cleavage
R R Adams et al.
Chromosoma, 110(2), 65-74 (2001-07-17)
The inner centromere protein (INCENP), which has previously been described in chicken, frog and mouse, is required for correct chromosome segregation and cytokinesis. We have identified the human INCENP gene by library screening and reverse transcription-polymerase chain reaction (RT-PCR) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service