Skip to Content
Merck
All Photos(1)

Key Documents

SRP5362

Sigma-Aldrich

LMTK3, GST tagged from mouse

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AATYK3, MGC90743

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€339.00

€339.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€339.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€339.00


Please contact Customer Service for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~75 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Lmtk3(381983)

General description

LMTK3, or Lemur tyrosine kinase 3, belongs to the LMR subfamily of tyr protein kinases. LMTK3 decreases the activity of protein kinase C (PKC) and also leads to phosphorylation of AKT (Ser473), thereby increasing binding of forkhead box O3 (FOXO3) to the ESR1 promoter. LMTK3 phosphorylates ER alpha, protecting it from proteasomal degradation in vitro. Knockdown of LMTK3 leads to a reduction of tumor volume in an orthotopic mouse model and abrogates proliferation of ER alpha+ but not ER alpha- cells, indicative of its role in ER alpha activity. These findings reveal LMTK3 as a new therapeutic target for breast cancer.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service