SRP5355
MYT1, GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
DKFZp547K1610, FLJ20093, PKMYT1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~93 kDa
NCBI accession no.
application(s)
cell analysis
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PKMYT1(9088)
General description
The PKMYT1 (protein kinase, membrane associated tyrosine/threonine 1) gene is differentially expressed between fetal liver and postnatal liver. This gene is mapped to human chromosome 16p13.3. It is also known to be localized to immature oocytes. MYT1 (myelin transcription factor 1) is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system. The recombinant MYT1 fragments containing either the upstream 2 zinc fingers or the downstream 4 zinc fingers bound the same cis regulatory element in the PLP1 promoter in vitro and DNA binding required Zn2+, but not other divalent cations.
Biochem/physiol Actions
The PKMYT1 (protein kinase, membrane associated tyrosine/threonine 1) gene expression is essential for disease progression of colorectal cancer and hepatocellular carcinoma. The action of Cdc2 (cyclin-dependent kinase 2), an essential molecule required for the induction of mitosis, is regulated by PKMYT1. PKMYT1 binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Preparation Note
After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
PKMYT1 promoted the growth and motility of hepatocellular carcinoma cells by activating beta-catenin/TCF signaling
Experimental Cell Research, 358(2), 209-216 (2017)
Myt1 protein kinase is essential for Golgi and ER assembly during mitotic exit
The Journal of Cell Biology, 181(1), 89-103 (2008)
Protein kinase, membrane-associated tyrosine/threonine 1 is associated with the progression of colorectal cancer
Oncology Reports, 39(6), 2829-2836 (2018)
Nature genetics, 47(3), 199-208 (2015-01-20)
Long noncoding RNAs (lncRNAs) are emerging as important regulators of tissue physiology and disease processes including cancer. To delineate genome-wide lncRNA expression, we curated 7,256 RNA sequencing (RNA-seq) libraries from tumors, normal tissues and cell lines comprising over 43 Tb
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service