Skip to Content
Merck
All Photos(2)

Key Documents

SRP5289

Sigma-Aldrich

VRK2, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

9.6-14.4 nmol/min·mg

mol wt

~66 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... VRK2(7444)

General description

VRK2 (also known as vaccinia related kinase 2) is a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. VRK2 is widely expressed in human tissues and highly expressed in actively dividing cells, such as those in testis, leukocytes, fetal liver and carcinomas. VRK2 can be used to phosphorylate casein and itself undergo autophosphorylation. VRK2 interacts specifically with Epstein-Barr virus BHRF1, a homologue of Bcl-2, and this interaction is involved in protecting cells from apoptosis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service