Skip to Content
Merck
All Photos(2)

Documents

SRP5246

Sigma-Aldrich

PAK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC130000, MGC130001, PAKα

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

63-85 nmol/min·mg

mol wt

~95 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PAK1(5058)

General description

p21 (RAC1) activated kinase 1 (PAK1) is a member of the p21-activated kinases (PAKs) which have been implicated in the regulation of cell morphology, motility and transformation. PAK1 belongs to the Group I PAKs which also includes PAK2 and PAK3. The gene encoding this protein is localized on human chromosome 11q13.

Biochem/physiol Actions

p21-activated kinases (PAKs) are serine/threonine kinases that are activated by and are effectors of small GTPases, cell division control protein 42 (CDC42) and Ras-related C3 botulinum toxin substrate (RAC). PAK1 is a key regulator of the actin cytoskeleton, adhesion and cell motility. Inactive dimeric PAK1 is mainly cytosolic and interaction with the activators Cdc42-GTP and Rac1-GTP stimulates the kinase at the sites of cellular protrusions forming adhesions to the extracellular matrix. The protein has a role in numerous cellular processes and has been shown to be expressed in squamous non-small cell lung carcinomas.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essential role of CIB1 in regulating PAK1 activation and cell migration.
Leisner TM
The Journal of Cell Biology, 170(3), 465-476 (2005)
Dynamics of the Rho-family small GTPases in actin regulation and motility.
Spiering D
Cell adhesion & migration, 5(2), 170-180 (2011)
Targeting p21-activated kinase 1 (PAK1) to induce apoptosis of tumor cells.
Ong CC
Proceedings of the National Academy of Sciences of the USA, 108(17), 7177-7182 (2011)
M C Parrini et al.
Biochemical Society transactions, 33(Pt 4), 646-648 (2005-07-27)
Pak1 (p21-activated kinase 1) is a key regulator of the actin cytoskeleton, adhesion and cell motility. Such biological roles require a tight spatial and kinetic control of its localization and activity. We summarize here the current knowledge on Pak1 dynamics
Zahara M Jaffer et al.
The international journal of biochemistry & cell biology, 34(7), 713-717 (2002-04-16)
The p21-activated kinases (Paks) are serine/threonine protein kinases that bind to and, in some cases, are stimulated by activated forms of the small GTPases, Cdc42 and Rac. With the recent discovery of several novel isoforms, Paks are now categorized into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service