Skip to Content
Merck
All Photos(2)

Key Documents

SRP5235

Sigma-Aldrich

MLK2 (1-446), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAP3K10, MST

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€352.00

€352.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€352.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€352.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

41-55 nmol/min·mg

mol wt

~76 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP3K10(4294)

General description

MLK2 or mixed-lineage kinase 2 is a member of the serine/threonine kinase family, which activates MAPK8 JNK and MKK4/SEK1. MLK2 itself can be phoshorylated, and activated by JNK kinases. MLK2 is also known as mitogen-activated protein kinase kinase kinase 10 (MAP3K10). MLK2 functions preferentially on the JNK signaling pathway which is involved in nerve growth factor (NGF) induced neuronal apoptosis. MLK2 was isolated as a cDNA fragment from MKN28 gastric cancer cell library using degenerate PCR. MLK2 is highly expressed in brain, skeletal muscle, and testis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Katoh et al.
Oncogene, 10(7), 1447-1451 (1995-04-06)
Protein kinases play a key role in cell growth regulation. We have isolated a cDNA fragment of the MST gene from the MKN28 gastric cancer cell line cDNA pool by degenerate polymerase chain reaction. MST-cDNAs were cloned from the human
D S Dorow et al.
European journal of biochemistry, 234(2), 492-500 (1995-12-01)
Protein kinases play pivotal roles in the control of many cellular processes. In a search for protein kinases expressed in human epithelial tumour cells, we discovered two members of a novel protein kinase family [Dorow, D. S., Devereux, L., Dietzsch

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service