Skip to Content
Merck
All Photos(1)

Key Documents

SRP5149

Sigma-Aldrich

UCHL1, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€320.00

€320.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€320.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

€320.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~25 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UCHL1(7345)

Biochem/physiol Actions

UCHL1 is a member of the peptidase C12 family and a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin to generate the ubiquitin monomer. UCHL1 is expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in UCHL1 may be associated with Parkinson disease. The ligase and hydrolase activities of UCHL1 may play roles in proteasomal protein degradation, a process critical for neuronal health. Inhibition of UCHL1 in mouse hippocampal slices reduces normal synaptic function and long-term potentiation. PKA-CREB pathway mediates the effects of UCHL1 on Amyloid-beta-induced synaptic dysfunction and Uchl1 also reversed the inhibition of CREB phosphorylation induced by Amyloid-beta.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yichin Liu et al.
Cell, 111(2), 209-218 (2002-11-01)
The assumption that each enzyme expresses a single enzymatic activity in vivo is challenged by the linkage of the neuronal enzyme ubiquitin C-terminal hydrolase-L1 (UCH-L1) to Parkinson's disease (PD). UCH-L1, especially those variants linked to higher susceptibility to PD, causes
Bing Gong et al.
Cell, 126(4), 775-788 (2006-08-23)
The neuronal ubiquitin/proteasomal pathway has been implicated in the pathogenesis of Alzheimer's disease (AD). We now show that a component of the pathway, ubiquitin C-terminal hydrolase L1 (Uch-L1), is required for normal synaptic and cognitive function. Transduction of Uch-L1 protein
Marta Brzozowska et al.
International journal of molecular sciences, 24(12) (2023-06-28)
Aspirin (ASA) is a popular nonsteroidal anti-inflammatory drug (NSAID), which exerts its therapeutic properties through the inhibition of cyclooxygenase (COX) isoform 2 (COX-2), while the inhibition of COX-1 by ASA results in the formation of gastrointestinal side effects. Due to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service