Skip to Content
Merck
All Photos(1)

Key Documents

SRP5148

Sigma-Aldrich

Tubulin, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

M40, MGC117247, MGC16435, OK/SW-cl.56, TUBB, TUBB5

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€318.00

€318.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
€318.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€318.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~76 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TUBB(203068)

General description

Tubulin-β 1 (TUBB1) protein is a major constituent of microtubules.[1] Previous work suggested that the "microtubule binding domain" of tau (composed of three or four imperfect 18-amino acid repeats, separated by 13- or 14-amino acid inter-repeat regions) can bind to the C-terminal ends of both α and β tubulin monomers. It is found in hematopoietic cells and platelet marginal bands.[2] The gene encoding TUBB1 is localized on human chromosome 6p21.[3]

Application

GST tagged tubulin has been used in Western blotting and in mass spec as a control.[4]

Biochem/physiol Actions

Tubulin-β 1 (TUBB1) interaction with microtubule-associated proteins (MAPs) such as tau is fundamental for microtubule structure and function. It has a role in the formation of the cytoskeleton.[1][4] Studies revealed that TUBB1 is the target of various antitubulin agents used in the treatment of cancer. Subsequent studies have also concluded that TUBB1 mutations in clinical samples are rare, and unlikely to contribute to drug resistance.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Chau et al.
Biochemistry, 37(51), 17692-17703 (1999-01-28)
The interaction between tubulin subunits and microtubule-associated proteins (MAPs) such as tau is fundamental for microtubule structure and function. Previous work has suggested that the "microtubule binding domain" of tau (composed of three or four imperfect 18-amino acid repeats, separated
The intricate relationship between microtubules and their associated motor proteins during axon growth and maintenance
Andreas Prokop
Neural Dev. (2013)
Root morphogenic pathways in Eucalyptus grandis are modified by the activity of protein arginine methyltransferases
Krista l
BMC plant biology (2017)
The beta 1 tubulin R307H single nucleotide polymorphism is associated with treatment failures in immune thrombocytopenia (ITP)
Paula A
British Journal of Haematology (2013)
An expressed beta-tubulin gene, TUBB, is located on the short arm of human chromosome 6 and two related sequences are dispersed on chromosomes 8 and 13.
Floyd_smith G
Experimental Cell Research (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service