Skip to Content
Merck
All Photos(2)

Key Documents

SRP5101

Sigma-Aldrich

ZAK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AZK, MLK7, MLT, MLTK, MRK, mlklak

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€486.00

€486.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€486.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€486.00


Please contact Customer Service for Availability

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

53-71 nmol/min·mg

mol wt

~82 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ZAK(51776)

General description

ZAK is a member of the MAPKKK family of signal transduction molecules and mediates gamma radiation signaling leading to cell cycle arrest. The activity of ZAK plays a role in cell cycle checkpoint regulation as well as being involved in regulating actin organization. Expression of kinase-dead ZAK in mouse fibroblasts disrupts actin stress fibers and causes morphologic changes. ZAK can activate JNK through MKK4/MKK7 and ERK5/p38-gamma via MKK3/MKK6. Expression of ZAK increases the population of cells in the G2/M phase of the cell cycle, whereas dominant-negative ZAK attenuated the G2 arrest caused by gamma radiation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaw-Ji Yang
Biochemical and biophysical research communications, 297(1), 105-110 (2002-09-11)
The leucine-zipper (LZ) and sterile-alpha motif (SAM) kinase (ZAK) belongs to the MAP kinase kinase kinase (MAP3K) when upon over-expression in mammalian cells activates the JNK/SAPK pathway. The mechanisms by which ZAK activity is regulated are not well understood. Co-expression
Eleanore A Gross et al.
The Journal of biological chemistry, 277(16), 13873-13882 (2002-02-12)
Mitogen-activated protein (MAP) kinase pathways are three-kinase modules that mediate diverse cellular processes and have been highly conserved among eukaryotes. By using a functional complementation screen in yeast, we have identified a human MAP kinase kinase kinase (MAPKKK) that shares

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service