Skip to Content
Merck
All Photos(2)

Key Documents

SRP5088

Sigma-Aldrich

TAK1-TAB1 (1-303), (437-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAP3K7, MGC57664, TAK1: MAP3K7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

8-12 nmol/min·mg

mol wt

~74 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

TAK1 is a serine/threonine protein kinase that mediates signaling by TGFβ and morphogenetic protein (BMP). In response to IL-1, TAK1 forms a kinase complex with TAB1 and this complex is required for the activation of nuclear factor kappa B (NfκB). TAK1 can also activate MAPK8/JNK and MAP2K4/MKK4 and thus play a role in the cell response to environmental stress. Tak1 is essential for thymocyte development and activation and deletion of TAK1 prevents maturation of single-positive thymocytes displaying CD4 or CD8. Thymocytes lacking TAK1 fail to activate NfκB and JNK and are prone to apoptosis upon stimulation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Hsing Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(31), 11677-11682 (2006-07-22)
The protein kinase TAK1 mediates the activation of NF-kappaB in response to stimulation by proinflammatory cytokines and microbial pathogens in the innate immunity pathways. However, the physiological function of TAK1 in the adaptive immunity pathways is unclear. By engineering mice
K Yamaguchi et al.
Science (New York, N.Y.), 270(5244), 2008-2011 (1995-12-22)
The mitogen-activated protein kinase (MAPK) pathway is a conserved eukaryotic signaling module that converts receptor signals into various outputs. MAPK is activated through phosphorylation by MAPK kinase (MAPKK), which is first activated by MAPKK kinase (MAPKKK). A genetic selection based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service