Skip to Content
Merck
All Photos(2)

Key Documents

SRP5073

Sigma-Aldrich

PTPN12 (1-355), active, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PTPG1, tcag7.1075

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€249.00

€249.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€249.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€249.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2520-3410 nmol/min·mg

mol wt

~66 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN12(5782)

General description

Protein tyrosine phosphatase-PEST (PTPN12), a ubiquitously expressed cytoplasmic tyrosine phosphatase, is thought to play an important role in cell adhesion and motility, cell migration, and signal transduction for antigen receptors in B and T lymphocytes. Signal transduction via tyrosine phosphorylation, normally fine-tuned by the concerted action of both protein tyrosine kinases and protein tyrosine phosphatases (PTPs), is a key mechanism in tumorigenesis. Studies suggest potential role for PTP-PEST in regulation of p130(cas) in mitogen- and cell adhesion-induced signaling events.

Physical form

Supplied in 20mM MOPS, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25mM DTT, 0.1mM PMSF, 30% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Garton et al.
Molecular and cellular biology, 16(11), 6408-6418 (1996-11-01)
PTP-PEST is a ubiquitously expressed, cytosolic, mammalian protein tyrosine phosphatase (PTP) which exhibits high specific activity in vitro. We have investigated the substrate specificity of PTP-PEST by a novel substrate-trapping approach in combination with in vitro dephosphorylation experiments. We initially
A Angers-Loustau et al.
The Journal of cell biology, 144(5), 1019-1031 (1999-03-23)
In this article, we show that, in transfected COS-1 cells, protein tyrosine phosphatase (PTP)-PEST translocates to the membrane periphery following stimulation by the extracellular matrix protein fibronectin. When plated on fibronectin, PTP-PEST (-/-) fibroblasts display a strong defect in motility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service