Skip to Content
Merck
All Photos(3)

Documents

SRP5011

Sigma-Aldrich

CDK5/p35., active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDKN5, PSSALRE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

157-213 nmol/min·mg

mol wt

~59 kDa (CDK5)
~60 kDa (p35)

technique(s)

activity assay: suitable

solubility

soluble
water: soluble

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Research area: Apoptosis

Cdk5 is a proline-directed serine/threonine protein kinase that belongs to the family of cyclin-dependent kinases (CDKs). Active CDK5, also known as neuronal cdc2-like kinase is a heterodimer of CDK5 and is essential for its kinase activity. They are found abundantly in the mammalian brain.

Application

CDK5 has been used as a component of a buffer during the measurement of the in vitro phosphorylation of peroxisome proliferator-activated receptor gamma (PPARγ).

Biochem/physiol Actions

CDK5 is a neuron-specific kinase that plays a vital role in the development, functioning, and diseases of the central nervous system. Its optimum functioning is highly essential for proper neuronal migration, differentiation, axonal elongation, and synaptic function. CDK5 modulates neurite outgrowth, and axonal growth by interacting with several substrates like N- cadherin, Rap Guanine Nucleotide Exchange Factor 2(RapGEF2), focal adhesion kinase (FAK), p21 activating kinase 1, serotonin 6 receptor and Ras-related protein 8. CDK5 also plays a key role in the formation of the synapse, its maintenance, and communication. It is involved in the formation and retraction of the dendritic spines. It also regulates neuron secretion at the synapse by phosphorylating key mediators such as Synaspsin1. In addition, CDK5, present in the synaptic membrane phosphorylates the erythroblastic leukemia viral oncogene homolog B receptor (ErbB), resulting in increased acetylcholine receptor transcription. Lastly, CDK5 is also involved in the regulation of various other cellular processes like cell cycle, cell survival, apoptosis, gene regulation, etc.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide Capelli et al.
Biomolecules, 13(4) (2023-05-16)
PPARγ represents a key target for the treatment of type 2 diabetes and metabolic syndrome. To avoid serious adverse effects related to the PPARγ agonism profile of traditional antidiabetic drugs, a new opportunity is represented by the development of molecules
D Tang et al.
Progress in cell cycle research, 2, 205-216 (1996-01-01)
While cyclin-dependent kinase 5 (Cdk5) is widely distributed in mammalian tissues and in cultured cell lines, Cdk5-associated kinase activity has been demonstrated only in mammalian brains. An active form of Cdk5, called neuronal cdc2-like kinase (Nclk) has been purified from
Franck Peiretti et al.
Journal of medicinal chemistry, 63(21), 13124-13139 (2020-11-04)
A proprietary library of novel N-aryl-substituted amino acid derivatives bearing a hydroxamate head group allowed the identification of compound 3a that possesses weak proadipogenic and peroxisome proliferator-activated receptor γ (PPARγ) activating properties. The systematic optimization of 3a, in order to
Feng He et al.
Cellular and molecular neurobiology, 40(6), 897-909 (2020-02-06)
A30P and A53T mutations in the gene encoding alpha-synuclein-a presynaptic protein-are the most frequently identified genetic causes of Parkinson's disease (PD). Aberrant alpha-synuclein likely plays central roles in dopaminergic neuronal death and motor symptoms in PD. This study investigated the
Antonio Laghezza et al.
Journal of medicinal chemistry, 61(18), 8282-8298 (2018-09-11)
A new series of derivatives of the PPARα/γ dual agonist 1 allowed us to identify the ligand ( S)-6 as a potent partial agonist of both PPARα and γ subtypes. X-ray studies in PPARγ revealed two different binding modes of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service