Skip to Content
Merck
All Photos(1)

Key Documents

SRP4304

Sigma-Aldrich

NGF-beta from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

Nerve Growth Factor β, Ngf, Ngfb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized

potency

0.2 ng/mL ED50

mol wt

~13.4 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ngf(18049)

General description

Nerve growth factor-β (NGF-β) belongs to the neurotrophin family.[1] The active form of human NGF-β is a homodimer, non-glycosylated chain.[2]

Biochem/physiol Actions

Nerve growth factor-β (NGF-β) is a potent neurotrophic factor that supports the growth and survivability of nerve and/or glial cells.[1] It brings pro-survival effects with the help of receptor tyrosine kinase TrkA.[3] NGF-β is associated with diabetic neuropathy and ischemic heart disease.[1] It is also involved in airway nerve system.[4] NGF-β is also responsible for the pain which is linked with osteoarthritis (OA).[5] It is involved in liver fibrosis, carcinogenesis, angiogenesis and cholangiocyte proliferation.[6]

Physical form

Lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL containing 1% HSA or BSA. The solution can be diluted into other aqueous buffers and stored at 4°C for 2-7 days or store at –20°C for future use.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Up-regulation of nerve growth factor in cholestatic livers and its hepatoprotective role against oxidative stress.
Tsai MS
PLoS ONE, 9, e112113-e112113 (2014)
Nerve growth factor rescues diabetic mice heart after ischemia/reperfusion injury via up-regulation of the TRPV1 receptor.
Zheng LR
The Journal of Diabetic Complications, 29, 323-328 (2015)
Nerve growth factor-mediated inhibition of apoptosis post-caspase activation is due to removal of active caspase-3 in a lysosome-dependent manner.
Mnich K
Cell Death & Disease, 5, e1202-e1202 (2014)
Induction of nerve growth factor expression and release by mechanical and inflammatory stimuli in chondrocytes: possible involvement in osteoarthritis pain.
Arthritis Research & Therapy, 16, R16-R16 (2014)
The Effects of Nerve Growth Factor on Nicotinic Synaptic Transmission in Mouse Airway Parasympathetic Neurons.
Weigand LA
American Journal of Respiratory Cell and Molecular Biology, 53, 443-449 (2015)

Questions

  1. Does Product No. SRP4304 contain any tags?

    1 answer
    1. The mouse NGF-beta product with Product No. SRP4304 does not contain any tags.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service