Skip to Content
Merck
All Photos(1)

Key Documents

SRP3081

Sigma-Aldrich

IL-17B human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

IL-20, NIRF

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€328.00

€328.00


Please contact Customer Service for Availability


Select a Size

Change View
25 μG
€328.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€328.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

36.6 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL17B(27190)

General description

IL-17B is a disulfide-linked homodimer of two 161 amino acid polypeptide chains. It belongs to the IL-17 family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17B is expressed by T-cells and has been shown to stimulate release of TNF-a and IL-1b from cells of the monocyte lineage. Recombinant human IL-17B is a 36.6 kDa non-disulfide-linked homodimer of two 161 amino acid polypeptide chains.

Biochem/physiol Actions

IL-17B is a disulfide-linked homodimer of two 161 amino acid polypeptide chains. Recombinant human IL-17B is a 36.6 kDa non-disulfide-linked homodimer of two 161 amino acid polypeptide chains.

Sequence

MQPRSPKSKR KGQGRPGPLA PGPHQVPLDL VSRMKPYARM EEYERNIEEM VAQLRNSSEL AQRKCEVNLQ LWMSNKRSLS PWGYSINHDP SRIPVDLPEA RCLCLGCVNP FTMQEDRSMV SVPVFSQVPV RRRLCPPPPR TGPCRQRAVM ETIAVGCTCI F

Physical form

Lyophilized from 1x PBS, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Fossiez et al.
The Journal of experimental medicine, 183(6), 2593-2603 (1996-06-01)
Analysis of the cDNA encoding murine interleukin (IL) 17 (cytotoxic T lymphocyte associated antigen 8) predicted a secreted protein sharing 57% amino acid identity with the protein predicted from ORF13, an open reading frame of Herpesvirus saimiri. Here we report
T A Moseley et al.
Cytokine & growth factor reviews, 14(2), 155-174 (2003-03-26)
Interleukin-17 (IL-17) is a pro-inflammatory cytokine secreted by activated T-cells. Recently discovered related molecules are forming a family of cytokines, the IL-17 family. The prototype member of the family has been designated IL-17A. Due to recent advances in the human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service