Skip to Content
Merck
All Photos(1)

Documents

SRP3037

Sigma-Aldrich

FAS Receptor human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Apo I, CD95, Fas Antigen, TNFRSF6, soluble Fas receptor (sFasR)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

10-15 μg/mL

mol wt

17.6 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FAS(355)

General description

The gene FAS (apoptosis-mediating surface antigen) is mapped to human chromosome 10q24.1. The encoded protein is a cell surface receptor. FAS is a type I membrane receptor. It belongs to the tumor necrosis factor family of surface receptors.

Biochem/physiol Actions

FAS (apoptosis-mediating surface antigen) is mainly responsible for apoptotic signaling. It interacts with the ligand, Fas ligand (FasL), to initiate signaling events leading to cell death. Disturbances in the expression of Fas are associated with cancer development. Mutation in the gene might be associated with coronary artery disease. FAS plays a significant role in diabetic retinopathy in diabetes mellitus patients. This is due to AGE (advanced glycation end-product)-associated apoptosis via Fas/FasL. FAS might also be involved with the pathogenesis of pulmonary edema in Plasmodium falciparum-infected malaria patients by regulating cell death in the lungs.

Sequence

MRLSSKSVNA QVTDINSKGL ELRKTVTTVE TQNLEGLHHD GQFCHKPCPP GERKARDCTV NGDEPDCVPC QEGKEYTDKA HFSSKCRRCR LCDEGHGLEV EINCTRTQNT KCRCKPNFFC NSTVCEHCDP CTKCEHGIIK ECTLTSNTKC KEEGSRS

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphisms of extrinsic death receptor apoptotic genes (FAS -670 G>A, FASL -844 T>C) in coronary artery disease.
Apoptosis (2016)
Quantitative assessment of the association between Fas/FasL gene polymorphism and susceptibility to esophageal carcinoma in a north Chinese population.
Zhang M
Cancer Medicine (2016)
Advanced glycation end-product (AGE) induces apoptosis in human retinal ARPE-19 cells via promoting mitochondrial dysfunction and activating the Fas-FasL signaling.
Wang P
Bioscience, Biotechnology, and Biochemistry (2016)
Enhanced expression of Fas and FasL modulates apoptosis in the lungs of severe P. falciparum malaria patients with pulmonary edema.
Punsawad C
International Journal of Clinical and Experimental Pathology (2015)
Apoptosis by death factor.
S Nagata
Cell, 88(3), 355-365 (1997-02-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service