Skip to Content
Merck
All Photos(2)

Key Documents

SRP0727

Sigma-Aldrich

EGFR (D746-750 C797S), Active

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€366.00

€366.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€366.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€366.00


Please contact Customer Service for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

78-110 nmol/min·mg (radiometric)

mol wt

~89-100 kDa

concentration

0.1 μg/μL

accession no.

NM_005228

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EGFR(1956)

Application

Useful for kinase assay.

Biochem/physiol Actions

Recombinant human EGFR (d746-750 C797S) (668-end) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. EGFR is the receptor for members of the EGF family and is a transmembrane glycoprotein that has tyrosine kinase activity. Binding of epidermal growth factor to EGFR induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation, differentiation, motility, and cell survival (1). Activation of EGFR triggers mitogenic signaling in gastrointestinal mucosa, and its expression is upregulated in colon cancers and most neoplasms (2). Activation of EGFR triggers activation of the ERK-signaling pathway in normal gastric epithelial and colon cancer cell lines. Inactivation of EGFR with selective inhibitors significantly reduces ERK2 activation, c-fos mRNA expression and cell proliferation.

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service