Skip to Content
Merck
All Photos(1)

Key Documents

SRP0620

Sigma-Aldrich

CD38, His-Tag (Human)

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€435.00

€435.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€435.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€435.00


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

31 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD38(952)

General description

Human CD38, also known as cluster of differentiation 38 and cyclic ADP ribose hydrolase with C-terminal His-tag, GenBank Accession No. NM_001775, a.a. 43-300 expressed in a HEK293 cell expression system. MW = 31 kDa. This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service