Skip to Content
Merck
All Photos(2)

Key Documents

SRP0435

Sigma-Aldrich

PDE7B active mouse

recombinant, expressed in baculovirus infected Sf9 cells, ≥65% (SDS-PAGE)

Synonym(s):

phosphodiesterase 7B

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€515.00

€515.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€515.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€515.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥65% (SDS-PAGE)

form

aqueous solution

mol wt

65 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... PDE7B(29863)

General description

Mouse PDE7B (GenBank Accession No. NM_013875) amino acids 108-446 (end) with N-terminal GST-tag, MW=65 kDa, expressed in a Baculovirus-infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

1 unit is defined as the amount of enzyme that will convert 1 pmole of 3?, 5?-cAMP to 5? AMP per min at 30°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Reyes-Irisarri et al.
Neuroscience, 132(4), 1173-1185 (2005-04-29)
cAMP plays an important role as second messenger molecule controlling multiple cellular processes in the brain. cAMP levels depend critically on the phosphodiesterases (PDE) activity, enzymes responsible for the clearance of intracellular cAMP. We have examined the regional distribution and
Lingzhi Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(49), 19532-19537 (2008-11-27)
Cyclic nucleotide phosphodiesterase (PDE) isoforms can influence disease pathogenesis and be novel therapeutic targets. Because lower cAMP levels may contribute to the decreased apoptosis that occurs in chronic lymphocytic leukemia (CLL), we assessed the expression levels of PDE isoforms in
C Gardner et al.
Biochemical and biophysical research communications, 272(1), 186-192 (2000-06-29)
We have identified and characterised a novel member of the PDE7 family of cyclic nucleotide phosphodiesterases (PDE), which we have designated PDE7B. Mouse and human full-length cDNAs were isolated encoding a protein of 446 and 450 amino acids, respectively. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service