SRP0428
PDE2A active rat
recombinant, expressed in baculovirus infected Sf9 cells, ≥56% (SDS-PAGE)
Synonym(s):
phosphodiesterase 2A, cGMP-stimulated
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
biological source
rat
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥56% (SDS-PAGE)
form
aqueous solution
mol wt
133 kDa
packaging
pkg of 5 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
rat ... PDE2A(81743)
General description
Rat PDE2A (GenBank Accession No. NM_00143847) full length with N-terminal His-GST-tag, MW=133 kDa, expressed in a Baculovirus infected Sf9 cell expression system
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The two GAF domains in phosphodiesterase 2A have distinct roles in dimerization and in cGMP binding.
Proceedings of the National Academy of Sciences of the United States of America, 99(20), 13260-13265 (2002-09-25)
Cyclic nucleotide phosphodiesterases (PDEs) regulate all pathways that use cGMP or cAMP as a second messenger. Five of the 11 PDE families have regulatory segments containing GAF domains, 3 of which are known to bind cGMP. In PDE2 binding of
Blood, 105(9), 3569-3576 (2005-01-15)
The pleiotropic cytokine tumor necrosis factor-alpha (TNF-alpha) and thrombin lead to increased endothelial permeability in sepsis. Numerous studies demonstrated the significance of intracellular cyclic nucleotides for the maintenance of endothelial barrier function. Actions of cyclic adenosine monophosphate (cAMP) and cyclic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service