Skip to Content
Merck
All Photos(1)

Key Documents

SRP0381

Sigma-Aldrich

EZH2 /EED/SUZ12/RbAp48/AEBP2 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€710.00

€710.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€710.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€710.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

48 kDa (RbBP5)
51 kDa (EED)
53 kDa (AEBP2)
86 kDa (EZH2)
87 kDa (SUZ12)

packaging

pkg of 20 μg

shipped in

dry ice

storage temp.

−70°C

General description

Mutant version of EZH2 5 member complex, with a Tyr- to-Ser mutation on amino acids 646 of the EZH2 protein (GenBank Accession No. NM_004456). This mutation is often cited in the scientific literature as Y641S in variant 3 (GenBank Accession No. NM_001203247).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan D Morin et al.
Nature genetics, 42(2), 181-185 (2010-01-19)
Follicular lymphoma (FL) and the GCB subtype of diffuse large B-cell lymphoma (DLBCL) derive from germinal center B cells. Targeted resequencing studies have revealed mutations in various genes encoding proteins in the NF-kappaB pathway that contribute to the activated B-cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service