Skip to Content
Merck
All Photos(1)

Key Documents

SRP0239

Sigma-Aldrich

EGFR Active human

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

Synonym(s):

ERBB1, Epidermal growth factor receptor, HER1, PIG61, Proto-oncogene c-ErbB-1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€453.00

€453.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€453.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€453.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

potency

≥0.14 units/mg

mol wt

87.3 kDa

packaging

pkg of 10 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EGFR(1956)

General description

Epidermal growth factor receptor (EGFR) is a transmembrane protein and a member of a family of receptor tyrosine kinases (RTK) called avian erythroblastosis oncogene (BErbB). It contains a ligand-binding domain in its extracellular region, one transmembrane domain, and a tyrosine kinase-containing domain in its cytosolic region.[1] EGFR is encoded by the gene mapped to human chromosome 7p11.2.[2]

Application

EGFR Active human has been used in:
  • photo-cross-linking assays[3]
  • StavroX cross-linking site identification[3]
  • in vitro kinase assay[4]

Biochem/physiol Actions

Epidermal growth factor receptor (EGFR) functions as a receptor for EGF-family of proteins, which are peptide growth factors. EGFR and ligand play a crucial role early embryonic development and in the renewal of stem cells in tissues such as the gut, liver and skin. This receptor-ligand system is also involved in the pathogenesis and progression of various types of carcinomas.[1] In cancer, EGFR pathways promotes cellular proliferation, angiogenesis, metastasis and apoptosis.[5] The microRNA miR-218-5p interacts with and controls EGFR, and in non-small cell lung cancer (NSCLC), the levels of EGFR are inversely related to miR-218-5p. miR-218-5p inhibits proliferation and migration of lung cancer cells by binding with EGFR, and thus, has potential in future NSCLC therapies.[6]

Unit Definition

One unit is defined as the amount of enzyme that will phosphorylate 1 pmol of Tyr substrate per minute at pH 7.4 and 30°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol, 10 mM glutathione and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marites Pasuelo Melancon et al.
ACS nano, 8(5), 4530-4538 (2014-04-24)
The purpose of this study was to compare the binding affinity and selective targeting of aptamer- and antibody-coated hollow gold nanospheres (HAuNS) targeted to epidermal growth factor receptors (EGFR). EGFR-targeting aptamers were conjugated to HAuNS (apt-HAuNS) by attaching a thiol-terminated
Julie K L Sinclair et al.
Biochemistry, 60(7), 500-512 (2021-02-10)
We previously reported a family of hydrocarbon-stapled peptides designed to interact with the epidermal growth factor receptor (EGFR) juxtamembrane (JM) segment, blocking its ability to form a coiled coil dimer that is essential for receptor activation. These hydrocarbon-stapled peptides, most
Filippo Lococo et al.
International journal of molecular sciences, 16(8), 19612-19630 (2015-08-22)
Assessment of biological diagnostic factors providing clinically-relevant information to guide physician decision-making are still needed for diseases with poor outcomes, such as non-small cell lung cancer (NSCLC). Epidermal growth factor receptor (EGFR) is a promising molecule in the clinical management
Tumor-suppressive miR-218-5p inhibits cancer cell proliferation and migration via EGFR in non-small cell lung cancer.
Zhu K, et al.
Oncotarget, 7(19), 28075-28085 (2016)
Chromosome 7p11.2 (EGFR) variation influences glioma risk.
Sanson M, et al.
Human Molecular Genetics, 20(14), 2897-2904 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service