Skip to Content
Merck
All Photos(2)

Key Documents

SRP0112

Sigma-Aldrich

HDAC-10 human

recombinant, expressed in baculovirus infected insect cells, ≥10% (SDS-PAGE)

Synonym(s):

HD10, Histone deacetylase 10

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€485.00

€485.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€485.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€485.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥10% (SDS-PAGE)

form

aqueous solution

mol wt

79 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC10(83933)

General description

Human HDAC10, amino acids 1-481, GenBank Accession No. NM_032019, with N-terminal GST tag, MW = 79 kDa, expressed in baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to deacetylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20 and 50% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service